Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGATGTTTGCCTTATGGGGTCA[A/G]TGGGACTGCAACAGCAATATGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALGAPB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RALGAPB - Ral GTPase activating protein non-catalytic beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282917.1 | Intron | NP_001269846.1 | ||||
NM_001282918.1 | Intron | NP_001269847.1 | ||||
NM_020336.3 | Intron | NP_065069.1 | ||||
XM_005260462.2 | Intron | XP_005260519.1 | ||||
XM_005260463.2 | Intron | XP_005260520.1 | ||||
XM_005260464.2 | Intron | XP_005260521.1 | ||||
XM_005260465.2 | Intron | XP_005260522.1 | ||||
XM_017027966.1 | Intron | XP_016883455.1 | ||||
XM_017027967.1 | Intron | XP_016883456.1 | ||||
XM_017027968.1 | Intron | XP_016883457.1 | ||||
XM_017027969.1 | Intron | XP_016883458.1 | ||||
XM_017027970.1 | Intron | XP_016883459.1 |