Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGTTCGGGAAAGTAGTCATGA[A/G]GATAAAGCTGGTGGCAGCCATGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606610 | ||||||||||||||||||||
Literature Links: |
NSFL1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NSFL1C - NSFL1 cofactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206736.1 | 2440 | Missense Mutation | CTC,TTC | L,F 341 | NP_001193665.1 | |
NM_016143.4 | 2440 | Missense Mutation | CTC,TTC | L,F 339 | NP_057227.2 | |
NM_018839.4 | 2440 | Missense Mutation | CTC,TTC | L,F 308 | NP_061327.2 | |
XM_006723592.2 | 2440 | Missense Mutation | CTC,TTC | L,F 230 | XP_006723655.1 | |
XM_006723593.2 | 2440 | Missense Mutation | CTC,TTC | L,F 230 | XP_006723656.1 | |
XM_006723594.2 | 2440 | Missense Mutation | CTC,TTC | L,F 228 | XP_006723657.1 | |
XM_011529300.1 | 2440 | Missense Mutation | CTC,TTC | L,F 344 | XP_011527602.1 | |
XM_011529301.1 | 2440 | Missense Mutation | CTC,TTC | L,F 342 | XP_011527603.1 | |
XM_011529302.2 | 2440 | Missense Mutation | CTC,TTC | L,F 291 | XP_011527604.1 | |
XM_011529304.1 | 2440 | Missense Mutation | CTC,TTC | L,F 288 | XP_011527606.1 | |
XM_011529305.2 | 2440 | Intron | XP_011527607.1 | |||
XM_017027956.1 | 2440 | Missense Mutation | CTC,TTC | L,F 289 | XP_016883445.1 | |
XM_017027957.1 | 2440 | Missense Mutation | CTC,TTC | L,F 290 | XP_016883446.1 | |
XM_017027958.1 | 2440 | Missense Mutation | CTC,TTC | L,F 254 | XP_016883447.1 |