Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCCATCACCCCTGGCTCTACCC[C/T]GTCCCGTGGCTGCTGTAGTCAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF16B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF16B - kinesin family member 16B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199865.1 | 3979 | Silent Mutation | NP_001186794.1 | |||
NM_001199866.1 | 3979 | Intron | NP_001186795.1 | |||
NM_024704.4 | 3979 | Silent Mutation | NP_078980.3 | |||
XM_005260750.3 | 3979 | Intron | XP_005260807.1 | |||
XM_005260751.4 | 3979 | Intron | XP_005260808.1 | |||
XM_005260752.3 | 3979 | Intron | XP_005260809.1 | |||
XM_005260753.3 | 3979 | Silent Mutation | XP_005260810.1 | |||
XM_005260754.3 | 3979 | Silent Mutation | XP_005260811.1 | |||
XM_005260755.3 | 3979 | Silent Mutation | XP_005260812.1 | |||
XM_006723588.3 | 3979 | Silent Mutation | XP_006723651.1 | |||
XM_017027926.1 | 3979 | Silent Mutation | XP_016883415.1 |