Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGGTAGACACTCAGGATCTCTG[C/T]GATGATGCTGGGGTCCTCCAAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614355 | ||||||||||||||||||||
Literature Links: |
ACSS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSS1 - acyl-CoA synthetase short-chain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252675.1 | 2094 | Missense Mutation | NP_001239604.1 | |||
NM_001252676.1 | 2094 | Missense Mutation | NP_001239605.1 | |||
NM_001252677.1 | 2094 | Intron | NP_001239606.1 | |||
NM_032501.3 | 2094 | Missense Mutation | NP_115890.2 | |||
XM_006723659.1 | 2094 | Missense Mutation | XP_006723722.1 | |||
XM_011529388.1 | 2094 | Missense Mutation | XP_011527690.1 |