Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTCAGCCCACGTGTCCTCCCGA[C/T]ATGAGGCCCATTTTTACGAGTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607167 MIM: 601242 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYNLRB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYNLRB1 - dynein light chain roadblock-type 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP1LC3A - microtubule associated protein 1 light chain 3 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032514.3 | Intron | NP_115903.1 | ||||
NM_181509.2 | Intron | NP_852610.1 | ||||
XM_011529083.2 | Intron | XP_011527385.1 | ||||
XM_011529084.2 | Intron | XP_011527386.1 | ||||
XM_011529085.2 | Intron | XP_011527387.1 |