Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAATTGAAGAGATCCCTATATGC[C/T]CTGTTTTCTCAGTTTGGTCATGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603520 | ||||||||||||||||||||
Literature Links: |
SNRPB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNRPB2 - small nuclear ribonucleoprotein polypeptide B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003092.4 | 223 | Silent Mutation | GCC,GCT | A,A 29 | NP_003083.1 | |
NM_198220.2 | 223 | Silent Mutation | GCC,GCT | A,A 29 | NP_937863.1 |