Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCAATTTTAATGACATAGTGAA[A/G]CAAGGGTACGTGAGGATCCGGAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608334 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOK5 - docking protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294161.1 | 505 | Silent Mutation | AAA,AAG | K,K 10 | NP_001281090.1 | |
NM_018431.4 | 505 | Silent Mutation | AAA,AAG | K,K 10 | NP_060901.2 | |
NM_177959.2 | 505 | Intron | NP_808874.1 | |||
XM_011528904.1 | 505 | Intron | XP_011527206.1 |