Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAACACAGATTGAGCTCTGTGTG[C/T]CAGGCACAGGGCTGGGCACCAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616191 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLGAP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLGAP4 - DLG associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042486.3 | Intron | NP_001035951.1 | ||||
NM_014902.5 | Intron | NP_055717.2 | ||||
NM_183006.3 | Intron | NP_892118.1 | ||||
XM_005260332.4 | Intron | XP_005260389.1 | ||||
XM_005260333.4 | Intron | XP_005260390.1 | ||||
XM_011528688.2 | Intron | XP_011526990.1 | ||||
XM_011528689.2 | Intron | XP_011526991.1 | ||||
XM_011528691.2 | Intron | XP_011526993.1 | ||||
XM_011528692.2 | Intron | XP_011526994.1 | ||||
XM_017027725.1 | Intron | XP_016883214.1 | ||||
XM_017027726.1 | Intron | XP_016883215.1 | ||||
XM_017027727.1 | Intron | XP_016883216.1 |