Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGTGGAGAGTCAGGCTGGCGAG[C/T]GGCTCGCTGTCCACTCGGCAATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607114 MIM: 600751 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM33 - ADAM metallopeptidase domain 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGLEC1 - sialic acid binding Ig like lectin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023068.3 | 6061 | Silent Mutation | CCA,CCG | P,P 1560 | NP_075556.1 | |
XM_006723610.3 | 6061 | Silent Mutation | CCA,CCG | P,P 1560 | XP_006723673.1 | |
XM_011529324.2 | 6061 | Silent Mutation | CCA,CCG | P,P 1560 | XP_011527626.1 | |
XM_011529325.2 | 6061 | Silent Mutation | CCA,CCG | P,P 1560 | XP_011527627.1 | |
XM_011529326.2 | 6061 | Silent Mutation | CCA,CCG | P,P 1560 | XP_011527628.1 | |
XM_011529327.2 | 6061 | Silent Mutation | CCA,CCG | P,P 1560 | XP_011527629.1 | |
XM_011529328.2 | 6061 | Silent Mutation | CCA,CCG | P,P 1560 | XP_011527630.1 | |
XM_011529329.1 | 6061 | Silent Mutation | CCA,CCG | P,P 1560 | XP_011527631.1 | |
XM_011529330.1 | 6061 | Silent Mutation | CCA,CCG | P,P 1474 | XP_011527632.1 | |
XM_011529331.1 | 6061 | Silent Mutation | CCA,CCG | P,P 1471 | XP_011527633.1 | |
XM_011529332.1 | 6061 | Silent Mutation | CCA,CCG | P,P 1357 | XP_011527634.1 | |
XM_011529333.2 | 6061 | Intron | XP_011527635.1 |