Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCCGACAGATCATTGCTGTGGT[A/C]CTGGGTGTCATTTGGGGAGTTTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606577 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C20orf24 - chromosome 20 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199534.1 | 359 | Silent Mutation | GTA,GTC | V,V 55 | NP_001186463.1 | |
NM_018840.4 | 359 | Silent Mutation | GTA,GTC | V,V 55 | NP_061328.1 | |
NM_199483.2 | 359 | Silent Mutation | GTA,GTC | V,V 55 | NP_955777.1 |
SLA2 - Src like adaptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGIF2-C20orf24 - TGIF2-C20orf24 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199535.1 | 359 | Silent Mutation | GTA,GTC | V,V 81 | NP_001186464.1 |