Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGAAAAGTGATGATAGAAGAGG[A/C]GATGAAAGAGGAGATGAAGGAAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611157 MIM: 606459 | ||||||||||||||||||||
Literature Links: |
MRGBP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRGBP - MRG/MORF4L binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018270.5 | 493 | Missense Mutation | GAG,GCG | E,A 125 | NP_060740.1 |
OGFR - opioid growth factor receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OGFR-AS1 - OGFR antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |