Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGATGGCCACCAGGTCGATCAG[C/T]GTCAGCGGGCTCCGCAGGAAGGCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603788 | ||||||||||||||||||||
Literature Links: |
KCNG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNG1 - potassium voltage-gated channel modifier subfamily G member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002237.3 | 1368 | Silent Mutation | ACA,ACG | T,T 302 | NP_002228.2 | |
XM_006723785.2 | 1368 | Silent Mutation | ACA,ACG | T,T 302 | XP_006723848.1 | |
XM_011528800.1 | 1368 | Silent Mutation | ACA,ACG | T,T 302 | XP_011527102.1 | |
XM_011528801.1 | 1368 | Silent Mutation | ACA,ACG | T,T 302 | XP_011527103.1 | |
XM_011528802.1 | 1368 | Silent Mutation | ACA,ACG | T,T 302 | XP_011527104.1 | |
XM_011528803.2 | 1368 | Silent Mutation | ACA,ACG | T,T 302 | XP_011527105.1 | |
XM_011528804.1 | 1368 | Silent Mutation | ACA,ACG | T,T 302 | XP_011527106.1 | |
XM_011528805.2 | 1368 | Silent Mutation | ACA,ACG | T,T 302 | XP_011527107.1 | |
XM_011528806.1 | 1368 | Silent Mutation | ACA,ACG | T,T 302 | XP_011527108.1 |