Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGCAGCCACCTCAATGGTGGCC[A/G]TGTGCTCCTCAGGCCCGGGGGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607451 | ||||||||||||||||||||
Literature Links: |
GMEB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GMEB2 - glucocorticoid modulatory element binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012384.4 | 1399 | Missense Mutation | ACG,ATG | T,M 515 | NP_036516.1 | |
XM_005260202.4 | 1399 | Missense Mutation | ACG,ATG | T,M 515 | XP_005260259.1 | |
XM_006723776.2 | 1399 | Missense Mutation | ACG,ATG | T,M 437 | XP_006723839.1 | |
XM_011528779.2 | 1399 | Missense Mutation | ACG,ATG | T,M 437 | XP_011527081.1 |