Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTGGGAAGCACTGGTCGAAGAG[C/T]GTGATGGACAAACAGTACGAGATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610996 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DTD1 - D-tyrosyl-tRNA deacylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318043.1 | 378 | Silent Mutation | AGC,AGT | S,S 66 | NP_001304972.1 | |
NM_080820.5 | 378 | Silent Mutation | AGC,AGT | S,S 66 | NP_543010.3 | |
XM_005260872.4 | 378 | Silent Mutation | AGC,AGT | S,S 22 | XP_005260929.1 |