Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCCTTCGTGACATGGTGCGGAA[A/G]GAGTACCCCAACCTGAGCACATCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTPAL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTPAL - alpha tocopherol transfer protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039199.2 | 337 | Silent Mutation | AAA,AAG | K,K 67 | NP_001034288.1 | |
NM_001261839.1 | 337 | Silent Mutation | AAA,AAG | K,K 67 | NP_001248768.1 | |
NM_024331.4 | 337 | Silent Mutation | AAA,AAG | K,K 67 | NP_077307.2 | |
XM_011529043.2 | 337 | Silent Mutation | AAA,AAG | K,K 67 | XP_011527345.1 | |
XM_011529044.2 | 337 | Silent Mutation | AAA,AAG | K,K 67 | XP_011527346.1 | |
XM_011529045.2 | 337 | Silent Mutation | AAA,AAG | K,K 67 | XP_011527347.1 | |
XM_017028063.1 | 337 | Silent Mutation | AAA,AAG | K,K 67 | XP_016883552.1 |