Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAGGTGGGGGGTGCAGATGGAG[C/T]GGGTGTAGCAGAGGGGAGCACTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616825 MIM: 606726 | ||||||||||||||||||||
Literature Links: |
NCOA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NCOA5 - nuclear receptor coactivator 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020967.2 | 1389 | Missense Mutation | ACT,GCT | T,A 420 | NP_066018.1 | |
XM_005260474.3 | 1389 | Missense Mutation | ACT,GCT | T,A 315 | XP_005260531.1 | |
XM_011528951.2 | 1389 | UTR 3 | XP_011527253.1 | |||
XM_017027988.1 | 1389 | UTR 3 | XP_016883477.1 |
SLC12A5 - solute carrier family 12 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |