Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAACGTATACAGTTTGGAAAAATA[G/T]ATATGTCAGGGTATATACAGATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603666 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985410 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107985410 - uncharacterized LOC107985410 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX16 - syntaxin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001433.2 | Intron | NP_001001433.1 | ||||
NM_001134772.2 | Intron | NP_001128244.1 | ||||
NM_001134773.2 | Intron | NP_001128245.1 | ||||
NM_001204868.1 | Intron | NP_001191797.1 | ||||
NM_003763.5 | Intron | NP_003754.2 |
STX16-NPEPL1 - STX16-NPEPL1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |