Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTAAAGCCCATGTCCCGTTTGGT[C/T]ATCTGGTAATCAGTGTCCAGGGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608958 MIM: 604932 | ||||||||||||||||||||
Literature Links: |
ADA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADA - adenosine deaminase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000022.3 | 1081 | Missense Mutation | ATA,ATG | I,M 310 | NP_000013.2 | |
NM_001322050.1 | 1081 | Missense Mutation | ATA,ATG | I,M 175 | NP_001308979.1 | |
NM_001322051.1 | 1081 | Missense Mutation | ATA,ATG | I,M 286 | NP_001308980.1 |
PKIG - protein kinase (cAMP-dependent, catalytic) inhibitor gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |