Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGACACCATCCAGCAGCATGGG[C/T]GTGCCGTGGTGCCGGTCTGCAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610702 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C20orf27 - chromosome 20 open reading frame 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039140.2 | 1124 | Silent Mutation | ACA,ACG | T,T 172 | NP_001034229.1 | |
NM_001258429.1 | 1124 | Silent Mutation | ACA,ACG | T,T 147 | NP_001245358.1 | |
NM_001258430.1 | 1124 | Silent Mutation | ACA,ACG | T,T 147 | NP_001245359.1 | |
XM_011529266.2 | 1124 | Silent Mutation | ACA,ACG | T,T 131 | XP_011527568.1 |
HSPA12B - heat shock protein family A (Hsp70) member 12B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |