Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGAGATCTTCACCTTGACCGCA[A/C]TGAAAGCCCGGGCCGGCTGGTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613942 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUN5 - Sad1 and UNC84 domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080675.3 | 1100 | Missense Mutation | AGT,ATT | S,I 336 | NP_542406.2 | |
XM_011528573.1 | 1100 | Missense Mutation | AGT,ATT | S,I 359 | XP_011526875.1 | |
XM_011528574.1 | 1100 | Missense Mutation | AGT,ATT | S,I 311 | XP_011526876.1 | |
XM_011528575.1 | 1100 | Missense Mutation | AGT,ATT | S,I 246 | XP_011526877.1 | |
XM_011528576.1 | 1100 | Intron | XP_011526878.1 |