Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCCGCCGGCCCTGGCCCTCACT[A/G]TGTTGGAAGGTAAGGCCAGATGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610666 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929937 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929937 - uncharacterized LOC101929937 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017028182.1 | 460 | Intron | XP_016883671.1 |
NRSN2 - neurensin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323679.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | NP_001310608.1 | |
NM_001323680.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | NP_001310609.1 | |
NM_001323681.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | NP_001310610.1 | |
NM_001323682.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | NP_001310611.1 | |
NM_001323683.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | NP_001310612.1 | |
NM_001323684.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | NP_001310613.1 | |
NM_001323685.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | NP_001310614.1 | |
NM_024958.2 | 460 | Silent Mutation | CTA,CTG | L,L 60 | NP_079234.1 | |
XM_006723630.3 | 460 | Silent Mutation | CTA,CTG | L,L 60 | XP_006723693.1 | |
XM_011529360.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | XP_011527662.1 | |
XM_011529362.2 | 460 | Silent Mutation | CTA,CTG | L,L 60 | XP_011527664.1 | |
XM_011529363.2 | 460 | Silent Mutation | CTA,CTG | L,L 60 | XP_011527665.1 | |
XM_017028073.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | XP_016883562.1 | |
XM_017028074.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | XP_016883563.1 | |
XM_017028075.1 | 460 | Silent Mutation | CTA,CTG | L,L 60 | XP_016883564.1 | |
XM_017028076.1 | 460 | Silent Mutation | CTA,CTG | L,L 152 | XP_016883565.1 |