Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTCTGTTTCCCTGATAAGTGCC[G/T]GAGTACCAGGGGGAGCCGGATGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616177 MIM: 147520 | ||||||||||||||||||||
Literature Links: |
DDRGK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDRGK1 - DDRGK domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITPA - inosine triphosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267623.1 | 321 | Intron | NP_001254552.1 | |||
NM_001324236.1 | 321 | UTR 5 | NP_001311165.1 | |||
NM_001324237.1 | 321 | UTR 5 | NP_001311166.1 | |||
NM_001324238.1 | 321 | UTR 5 | NP_001311167.1 | |||
NM_001324240.1 | 321 | Silent Mutation | CCG,CCT | P,P 43 | NP_001311169.1 | |
NM_033453.3 | 321 | Silent Mutation | CCG,CCT | P,P 43 | NP_258412.1 | |
NM_181493.3 | 321 | Silent Mutation | CCG,CCT | P,P 26 | NP_852470.1 | |
XM_006723564.3 | 321 | Silent Mutation | CCG,CCT | P,P 43 | XP_006723627.1 | |
XM_006723565.3 | 321 | Intron | XP_006723628.1 | |||
XM_011529234.2 | 321 | Silent Mutation | CCG,CCT | P,P 43 | XP_011527536.1 |