Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTACCTTGCTACCAGGGAGGAGAT[C/T]TGTAGGATGGTGCAGGTGAAGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605273 MIM: 606577 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDRG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDRG3 - NDRG family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022477.3 | Intron | NP_071922.2 | ||||
NM_032013.3 | Intron | NP_114402.1 | ||||
XM_006723837.3 | Intron | XP_006723900.1 | ||||
XM_006723838.3 | Intron | XP_006723901.1 | ||||
XM_011528925.2 | Intron | XP_011527227.1 | ||||
XM_011528926.2 | Intron | XP_011527228.1 | ||||
XM_011528927.2 | Intron | XP_011527229.1 | ||||
XM_011528928.2 | Intron | XP_011527230.1 | ||||
XM_017027978.1 | Intron | XP_016883467.1 | ||||
XM_017027979.1 | Intron | XP_016883468.1 | ||||
XM_017027980.1 | Intron | XP_016883469.1 |
SLA2 - Src like adaptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |