Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGGACCCTGCTCTTCCGCAGCC[A/G]CAGCCGCCTGACGCACCTATCCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116949 | ||||||||||||||||||||
Literature Links: |
CDC25B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC25B - cell division cycle 25B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287516.1 | 996 | Missense Mutation | CAC,CGC | H,R 18 | NP_001274445.1 | |
NM_001287517.1 | 996 | Missense Mutation | CAC,CGC | H,R 4 | NP_001274446.1 | |
NM_001287518.1 | 996 | Missense Mutation | CAC,CGC | H,R 18 | NP_001274447.1 | |
NM_001287519.1 | 996 | UTR 5 | NP_001274448.1 | |||
NM_001287520.1 | 996 | UTR 5 | NP_001274449.1 | |||
NM_001287522.1 | 996 | UTR 5 | NP_001274451.1 | |||
NM_001287524.1 | 996 | Intron | NP_001274453.1 | |||
NM_004358.4 | 996 | Missense Mutation | CAC,CGC | H,R 68 | NP_004349.1 | |
NM_021872.3 | 996 | Missense Mutation | CAC,CGC | H,R 82 | NP_068658.1 | |
NM_021873.3 | 996 | Missense Mutation | CAC,CGC | H,R 82 | NP_068659.1 |