Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGTTTCACATTTTCCCTCCCCA[C/G]CCATGTAAAGCTTGTCCCAAAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF343 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF343 - zinc finger protein 343 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282495.1 | 3169 | UTR 3 | NP_001269424.1 | |||
NM_001282496.1 | 3169 | UTR 3 | NP_001269425.1 | |||
NM_001282497.1 | 3169 | UTR 3 | NP_001269426.1 | |||
NM_001282498.1 | 3169 | UTR 3 | NP_001269427.1 | |||
NM_001282499.1 | 3169 | Intron | NP_001269428.1 | |||
NM_001321800.1 | 3169 | UTR 3 | NP_001308729.1 | |||
NM_001321801.1 | 3169 | UTR 3 | NP_001308730.1 | |||
NM_001321802.1 | 3169 | UTR 3 | NP_001308731.1 | |||
NM_001321803.1 | 3169 | UTR 3 | NP_001308732.1 | |||
NM_001321805.1 | 3169 | UTR 3 | NP_001308734.1 | |||
NM_024325.5 | 3169 | UTR 3 | NP_077301.4 | |||
XM_017028062.1 | 3169 | UTR 3 | XP_016883551.1 |