Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGCCAGAAATGCTTATCCTCAT[G/T]TAGGACTGGAGGCAGCAGGACTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607650 MIM: 615997 | ||||||||||||||||||||
Literature Links: |
DEFB118 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DEFB118 - defensin beta 118 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DEFB119 - defensin beta 119 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271209.1 | 346 | Intron | NP_001258138.1 | |||
NM_153289.3 | 346 | Nonsense Mutation | TAA,TAC | *,Y 60 | NP_695021.2 | |
NM_153323.4 | 346 | Intron | NP_697018.1 |