Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCCTTATGGTTAGTATTAAGA[A/G]GAGGCCAGGTTTAAGCACAGGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALY PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RALY - RALY heterogeneous nuclear ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007367.3 | Intron | NP_031393.2 | ||||
NM_016732.2 | Intron | NP_057951.1 | ||||
XM_005260334.4 | Intron | XP_005260391.1 | ||||
XM_005260336.4 | Intron | XP_005260393.1 | ||||
XM_011528694.2 | Intron | XP_011526996.1 | ||||
XM_011528695.2 | Intron | XP_011526997.1 | ||||
XM_017027731.1 | Intron | XP_016883220.1 |
RALY-AS1 - RALY antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |