Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAACCTGGACAATATCTCTTTAGC[C/T]TCCTGCTGGTAGTGAAGCAAGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DSN1 - DSN1 homolog, MIS12 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145315.1 | 1060 | Silent Mutation | GAA,GAG | E,E 235 | NP_001138787.1 | |
NM_001145316.1 | 1060 | Silent Mutation | GAA,GAG | E,E 235 | NP_001138788.1 | |
NM_001145317.1 | 1060 | Silent Mutation | GAA,GAG | E,E 128 | NP_001138789.1 | |
NM_001145318.1 | 1060 | Silent Mutation | GAA,GAG | E,E 219 | NP_001138790.1 | |
NM_024918.3 | 1060 | Silent Mutation | GAA,GAG | E,E 235 | NP_079194.3 | |
XM_006723876.2 | 1060 | Silent Mutation | GAA,GAG | E,E 128 | XP_006723939.1 | |
XM_017028070.1 | 1060 | Silent Mutation | GAA,GAG | E,E 223 | XP_016883559.1 | |
XM_017028071.1 | 1060 | Silent Mutation | GAA,GAG | E,E 223 | XP_016883560.1 | |
XM_017028072.1 | 1060 | Silent Mutation | GAA,GAG | E,E 207 | XP_016883561.1 |