Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCACCTCTTTTACTACTTCGGC[C/T]GAGCCCACCTCCCCGGCGTGGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608958 MIM: 604932 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADA - adenosine deaminase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000022.3 | 811 | Silent Mutation | NP_000013.2 | |||
NM_001322050.1 | 811 | Silent Mutation | NP_001308979.1 | |||
NM_001322051.1 | 811 | Intron | NP_001308980.1 |
PKIG - protein kinase (cAMP-dependent, catalytic) inhibitor gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |