Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTGATTATTGCATTAGAGTCTA[C/T]CTTCCCTACTAGTCGGTAAGCTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602905 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNS1 - potassium voltage-gated channel modifier subfamily S member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322799.1 | 3901 | UTR 3 | NP_001309728.1 | |||
NM_002251.4 | 3901 | UTR 3 | NP_002242.2 | |||
XM_017027846.1 | 3901 | UTR 3 | XP_016883335.1 |