Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCTTCAGAGAGCAGAGGTGAAT[A/G]CTTTCTCATGAAGAAACGCCAACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614272 | ||||||||||||||||||||
Literature Links: |
FASTKD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FASTKD5 - FAST kinase domains 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021826.4 | 2587 | Missense Mutation | GCA,GTA | A,V 759 | NP_068598.1 |
UBOX5 - U-box domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267584.1 | 2587 | Intron | NP_001254513.1 | |||
NM_014948.3 | 2587 | Intron | NP_055763.1 | |||
NM_199415.2 | 2587 | Intron | NP_955447.1 |
UBOX5-AS1 - UBOX5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |