Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATTTGTTTACATTTCTCTGGA[C/T]TGGGAATGGCACTATATACAGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615566 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTBD3 - BTB domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282550.1 | Intron | NP_001269479.1 | ||||
NM_001282551.1 | Intron | NP_001269480.1 | ||||
NM_001282552.2 | Intron | NP_001269481.1 | ||||
NM_001282554.1 | Intron | NP_001269483.1 | ||||
NM_014962.3 | Intron | NP_055777.1 | ||||
NM_181443.3 | Intron | NP_852108.2 | ||||
XM_017027728.1 | Intron | XP_016883217.1 | ||||
XM_017027729.1 | Intron | XP_016883218.1 | ||||
XM_017027730.1 | Intron | XP_016883219.1 |