Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCCCTTCTGCACAGAACTCAAC[C/G]CTGAGGAGCAGGCCCTGAAGGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607976 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX4I2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX4I2 - cytochrome c oxidase subunit 4I2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032609.2 | 250 | Missense Mutation | CCT,GCT | P,A 59 | NP_115998.2 | |
XM_005260579.4 | 250 | Missense Mutation | CCT,GCT | P,A 64 | XP_005260636.1 | |
XM_005260580.4 | 250 | Missense Mutation | CCT,GCT | P,A 64 | XP_005260637.1 | |
XM_005260581.3 | 250 | Missense Mutation | CCT,GCT | P,A 59 | XP_005260638.1 |