Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCGTGTAGCTGCGCCTGTAGGT[C/T]TTGGGGTCGCGGGCGCGGCTCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600288 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXA2 - forkhead box A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021784.4 | 637 | Silent Mutation | AAA,AAG | K,K 155 | NP_068556.2 | |
NM_153675.2 | 637 | Silent Mutation | AAA,AAG | K,K 149 | NP_710141.1 |
LINC00261 - long intergenic non-protein coding RNA 261 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC01384 - long intergenic non-protein coding RNA 1384 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |