Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCTGGTGAACCAGCACAGCAT[C/G]GTGAGTGTGTCCACGCCCATCGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601242 MIM: 608528 | ||||||||||||||||||||
Literature Links: |
MAP1LC3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP1LC3A - microtubule associated protein 1 light chain 3 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032514.3 | 737 | Missense Mutation | ATC,ATG | I,M 88 | NP_115903.1 | |
NM_181509.2 | 737 | Missense Mutation | ATC,ATG | I,M 92 | NP_852610.1 | |
XM_011529083.2 | 737 | Missense Mutation | ATC,ATG | I,M 190 | XP_011527385.1 | |
XM_011529084.2 | 737 | Missense Mutation | ATC,ATG | I,M 190 | XP_011527386.1 | |
XM_011529085.2 | 737 | Missense Mutation | ATC,ATG | I,M 149 | XP_011527387.1 |
PIGU - phosphatidylinositol glycan anchor biosynthesis class U | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080476.4 | 737 | Intron | NP_536724.1 | |||
XM_011528542.2 | 737 | Intron | XP_011526844.1 | |||
XM_017027664.1 | 737 | Intron | XP_016883153.1 | |||
XM_017027665.1 | 737 | Intron | XP_016883154.1 |