Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTGAACAAGTGGGAAGTGTACA[C/T]TGCCTGAAACCGTGCATCGTTAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ESF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESF1 - ESF1 nucleolar pre-rRNA processing protein homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276380.1 | 3853 | Missense Mutation | ATG,GTG | M,V 765 | NP_001263309.1 | |
NM_016649.3 | 3853 | Missense Mutation | ATG,GTG | M,V 765 | NP_057733.2 | |
XM_017027874.1 | 3853 | Missense Mutation | ATG,GTG | M,V 765 | XP_016883363.1 |