Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTAAAGGACACAAAATAACT[C/T]TAAAACAATCTATCCATTTCAGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131340 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC727993 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC727993 - uncharacterized LOC727993 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDYN - prodynorphin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190892.1 | 2483 | UTR 3 | NP_001177821.1 | |||
NM_001190898.2 | 2483 | UTR 3 | NP_001177827.1 | |||
NM_001190899.2 | 2483 | UTR 3 | NP_001177828.1 | |||
NM_001190900.1 | 2483 | UTR 3 | NP_001177829.1 | |||
NM_024411.4 | 2483 | UTR 3 | NP_077722.1 | |||
XM_011529244.1 | 2483 | UTR 3 | XP_011527546.1 | |||
XM_011529245.1 | 2483 | UTR 3 | XP_011527547.1 | |||
XM_011529246.2 | 2483 | UTR 3 | XP_011527548.1 | |||
XM_011529247.1 | 2483 | UTR 3 | XP_011527549.1 | |||
XM_011529248.1 | 2483 | UTR 3 | XP_011527550.1 | |||
XM_011529249.2 | 2483 | UTR 3 | XP_011527551.1 | |||
XM_011529250.2 | 2483 | UTR 3 | XP_011527552.1 | |||
XM_017027878.1 | 2483 | UTR 3 | XP_016883367.1 |