Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACTCGAGCCTCACGGGGGATGG[C/T]GTATGCTGTCACCACCAGAAGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMF1 - proteasome inhibitor subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM74B - transmembrane protein 74B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304748.1 | 1277 | Missense Mutation | ACC,GCC | T,A 142 | NP_001291677.1 | |
NM_001304749.1 | 1277 | Missense Mutation | ACC,GCC | T,A 118 | NP_001291678.1 | |
NM_018354.2 | 1277 | Missense Mutation | ACC,GCC | T,A 142 | NP_060824.1 | |
XM_011529281.2 | 1277 | Missense Mutation | ACC,GCC | T,A 199 | XP_011527583.1 | |
XM_011529282.2 | 1277 | Missense Mutation | ACC,GCC | T,A 142 | XP_011527584.1 | |
XM_011529283.2 | 1277 | Missense Mutation | ACC,GCC | T,A 142 | XP_011527585.1 | |
XM_011529284.2 | 1277 | Missense Mutation | ACC,GCC | T,A 142 | XP_011527586.1 | |
XM_011529285.2 | 1277 | Missense Mutation | ACC,GCC | T,A 118 | XP_011527587.1 | |
XM_017027923.1 | 1277 | Missense Mutation | ACC,GCC | T,A 174 | XP_016883412.1 | |
XM_017027924.1 | 1277 | Missense Mutation | ACC,GCC | T,A 142 | XP_016883413.1 | |
XM_017027925.1 | 1277 | Missense Mutation | ACC,GCC | T,A 118 | XP_016883414.1 |