Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAATATTACTTTCTACTTCATAG[A/C]ACTATTGTGAAGATTAATTGAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603908 MIM: 614663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2S2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2S2 - eukaryotic translation initiation factor 2 subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316363.1 | Intron | NP_001303292.1 | ||||
NM_001316364.1 | Intron | NP_001303293.1 | ||||
NM_003908.4 | Intron | NP_003899.2 | ||||
XM_017028118.1 | Intron | XP_016883607.1 |
RALY - RALY heterogeneous nuclear ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |