Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGGGGCCCAGGAGGACAGCTAT[A/G]AGGAGCAGCGCAGTAAGGGATGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603889 MIM: 605466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIRPB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIRPB1 - signal regulatory protein beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRPG - signal regulatory protein gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039508.1 | 845 | Silent Mutation | NP_001034597.1 | |||
NM_018556.3 | 845 | Silent Mutation | NP_061026.2 | |||
NM_080816.2 | 845 | Silent Mutation | NP_543006.2 | |||
XM_005260749.3 | 845 | Intron | XP_005260806.1 | |||
XM_011529286.2 | 845 | Silent Mutation | XP_011527588.1 | |||
XM_011529287.2 | 845 | Intron | XP_011527589.1 |
SIRPG-AS1 - SIRPG antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |