Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCCATTCCAGTATTACCTCTTT[A/G]TACTGACAGTAGACCACAAACGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608187 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCM8 - minichromosome maintenance 8 homologous recombination repair factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT6 - tRNA methyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281467.1 | 1458 | Silent Mutation | TAC,TAT | Y,Y 233 | NP_001268396.1 | |
NM_015939.4 | 1458 | Silent Mutation | TAC,TAT | Y,Y 403 | NP_057023.2 | |
XM_017027875.1 | 1458 | Silent Mutation | TAC,TAT | Y,Y 221 | XP_016883364.1 |