Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAAACTGTACAATGAGAAAAGC[C/T]ACTTATGATTTTTTGTTGTTGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH26 - cadherin 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021810.4 | Intron | NP_068582.2 | ||||
NM_177980.2 | Intron | NP_817089.1 | ||||
XM_011528969.2 | Intron | XP_011527271.1 | ||||
XM_011528970.2 | Intron | XP_011527272.1 | ||||
XM_017027994.1 | Intron | XP_016883483.1 | ||||
XM_017027995.1 | Intron | XP_016883484.1 | ||||
XM_017027996.1 | Intron | XP_016883485.1 | ||||
XM_017027997.1 | Intron | XP_016883486.1 | ||||
XM_017027998.1 | Intron | XP_016883487.1 | ||||
XM_017027999.1 | Intron | XP_016883488.1 | ||||
XM_017028000.1 | Intron | XP_016883489.1 | ||||
XM_017028001.1 | Intron | XP_016883490.1 | ||||
XM_017028002.1 | Intron | XP_016883491.1 |