Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCACGGCGGCTGGGGGAGCCCTG[C/T]GACCAACTCCACGTCTGCGACGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603399 | ||||||||||||||||||||
Literature Links: |
KCNK15-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNK15-AS1 - KCNK15 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WISP2 - WNT1 inducible signaling pathway protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323369.1 | 387 | Silent Mutation | TGC,TGT | C,C 64 | NP_001310298.1 | |
NM_001323370.1 | 387 | Silent Mutation | TGC,TGT | C,C 64 | NP_001310299.1 | |
NM_003881.3 | 387 | Silent Mutation | TGC,TGT | C,C 64 | NP_003872.1 | |
XM_017028116.1 | 387 | UTR 5 | XP_016883605.1 | |||
XM_017028117.1 | 387 | Silent Mutation | TGC,TGT | C,C 122 | XP_016883606.1 |