Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCTCCTGCATCTCATCAAACT[C/T]GGTAGATTGTGAACACAAATCCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603072 MIM: 600369 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AURKA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AURKA - aurora kinase A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CSTF1 - cleavage stimulation factor subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033521.1 | 368 | Silent Mutation | CTC,CTT | L,L 56 | NP_001028693.1 | |
NM_001033522.1 | 368 | Silent Mutation | CTC,CTT | L,L 56 | NP_001028694.1 | |
NM_001324.2 | 368 | Silent Mutation | CTC,CTT | L,L 56 | NP_001315.1 | |
XM_011528600.1 | 368 | Silent Mutation | CTC,CTT | L,L 56 | XP_011526902.1 |