Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTCAAACACATGAAAGGCTGCA[G/T]GAATCACCTCGGGATCCTGTTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPIN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LPIN3 - lipin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301860.1 | Intron | NP_001288789.1 | ||||
XM_005260516.2 | Intron | XP_005260573.1 | ||||
XM_006723863.2 | Intron | XP_006723926.1 | ||||
XM_011528996.1 | Intron | XP_011527298.1 | ||||
XM_011528997.2 | Intron | XP_011527299.1 | ||||
XM_011528998.2 | Intron | XP_011527300.1 | ||||
XM_011528999.2 | Intron | XP_011527301.1 | ||||
XM_011529000.1 | Intron | XP_011527302.1 | ||||
XM_011529001.1 | Intron | XP_011527303.1 | ||||
XM_011529002.2 | Intron | XP_011527304.1 | ||||
XM_011529003.1 | Intron | XP_011527305.1 | ||||
XM_011529004.1 | Intron | XP_011527306.1 | ||||
XM_011529005.2 | Intron | XP_011527307.1 | ||||
XM_011529006.2 | Intron | XP_011527308.1 | ||||
XM_017028020.1 | Intron | XP_016883509.1 |