Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGCAACTGAAGCCTGAGGTCTC[C/T]CCAACGCAGGACATCAGGTGAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610924 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBCK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBCK1 - RANBP2-type and C3HC4-type zinc finger containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323956.1 | 654 | UTR 5 | NP_001310885.1 | |||
NM_001323958.1 | 654 | Intron | NP_001310887.1 | |||
NM_001323960.1 | 654 | Silent Mutation | TCC,TCT | S,S 50 | NP_001310889.1 | |
NM_006462.5 | 654 | Intron | NP_006453.1 | |||
NM_031229.3 | 654 | Silent Mutation | TCC,TCT | S,S 50 | NP_112506.2 | |
XM_005260645.1 | 654 | Silent Mutation | TCC,TCT | S,S 67 | XP_005260702.1 | |
XM_011529137.1 | 654 | Silent Mutation | TCC,TCT | S,S 67 | XP_011527439.1 | |
XM_011529138.1 | 654 | Silent Mutation | TCC,TCT | S,S 50 | XP_011527440.1 | |
XM_011529139.2 | 654 | Silent Mutation | TCC,TCT | S,S 67 | XP_011527441.1 | |
XM_011529140.1 | 654 | Silent Mutation | TCC,TCT | S,S 67 | XP_011527442.1 | |
XM_017027594.1 | 654 | UTR 5 | XP_016883083.1 | |||
XM_017027595.1 | 654 | UTR 5 | XP_016883084.1 |