Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTAGGCTTGGCACTGCTGCACCA[C/T]CAAAGGCAAACTTCATTGAAGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605371 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGEF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFGEF2 - ADP ribosylation factor guanine nucleotide exchange factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006420.2 | 324 | Missense Mutation | CCA,TCA | P,S 58 | NP_006411.2 | |
XM_005260252.3 | 324 | Missense Mutation | CCA,TCA | P,S 58 | XP_005260309.1 |