Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGACTTTTCTTTTGGGTAGTTTG[C/T]GGATTACTGATTTAATTGCTTTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608851 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XRN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XRN2 - 5'-3' exoribonuclease 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317960.1 | Intron | NP_001304889.1 | ||||
NM_012255.4 | Intron | NP_036387.2 | ||||
XM_017027722.1 | Intron | XP_016883211.1 | ||||
XM_017027723.1 | Intron | XP_016883212.1 | ||||
XM_017027724.1 | Intron | XP_016883213.1 |