Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCGCGCCCGGCCCCTCAGCGCCC[C/T]CAGCACGCCGCCGAGTCCCGCTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606145 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC2A10 - solute carrier family 2 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030777.3 | 64 | UTR 5 | NP_110404.1 | |||
XM_011529060.2 | 64 | Intron | XP_011527362.1 | |||
XM_011529061.2 | 64 | Intron | XP_011527363.1 | |||
XM_011529062.2 | 64 | Intron | XP_011527364.1 | |||
XM_011529063.2 | 64 | Intron | XP_011527365.1 | |||
XM_011529064.2 | 64 | Intron | XP_011527366.1 | |||
XM_011529065.2 | 64 | Intron | XP_011527367.1 | |||
XM_017028087.1 | 64 | UTR 5 | XP_016883576.1 |