Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCGCCCAGCTCCTCCGCCTCCC[C/G]TCGTGGGTCCTCAGGGTCCTCCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609809 MIM: 609493 MIM: 614639 | ||||||||||||||||||||
Literature Links: |
LIME1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIME1 - Lck interacting transmembrane adaptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC2A4RG - SLC2A4 regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB46 - zinc finger and BTB domain containing 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025224.3 | 1908 | Missense Mutation | CGG,GGG | R,G 543 | NP_079500.2 | |
XM_005260195.4 | 1908 | Missense Mutation | CGG,GGG | R,G 543 | XP_005260252.1 | |
XM_005260196.3 | 1908 | Missense Mutation | CGG,GGG | R,G 543 | XP_005260253.1 | |
XM_005260197.4 | 1908 | Missense Mutation | CGG,GGG | R,G 543 | XP_005260254.1 | |
XM_005260198.4 | 1908 | Missense Mutation | CGG,GGG | R,G 543 | XP_005260255.1 | |
XM_006723700.3 | 1908 | Missense Mutation | CGG,GGG | R,G 543 | XP_006723763.1 | |
XM_011528548.2 | 1908 | Missense Mutation | CGG,GGG | R,G 543 | XP_011526850.1 | |
XM_011528549.2 | 1908 | Intron | XP_011526851.1 | |||
XM_017027667.1 | 1908 | Intron | XP_016883156.1 |